Skip to Content
Merck
All Photos(1)

Key Documents

453455

Sigma-Aldrich

(R)-(+)-Glycidyl trityl ether

98%, optical purity ee: 98% (GLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H20O2
CAS Number:
Molecular Weight:
316.39
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

optical activity

[α]20/D +10.5°, c = 1 in chloroform

optical purity

ee: 98% (GLC)

mp

99-102 °C (lit.)

SMILES string

C1O[C@H]1COC(c2ccccc2)(c3ccccc3)c4ccccc4

InChI

1S/C22H20O2/c1-4-10-18(11-5-1)22(24-17-21-16-23-21,19-12-6-2-7-13-19)20-14-8-3-9-15-20/h1-15,21H,16-17H2/t21-/m1/s1

InChI key

XFSXUCMYFWZRAF-OAQYLSRUSA-N

Application

(R)-(+)-Glycidyl trityl ether may be used in the preparation of (R)-trityl(thiiranylmethyl)ether by reacting with thiourea. It may also be used as a starting material in the multi-step synthesis of (+)-actisonitrile.
Nucleotides with activity against herpes simplex virus 1 and 2, lipid ammonium salts, and thio analogs of phospholipids have been prepared recently through ring-opening of this epoxide with cytosine derivatives, amines, and thiols, respectively.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lin, H.-K. Gelb, M.H.
Journal of the American Chemical Society, 115, 3932-3932 (1993)
Bennett, M.J. et al.
Tetrahedron Letters, 36, 2207-2207 (1995)
Brodfuehrer, P.R. et al.
Tetrahedron Letters, 35, 3243-3243 (1994)
Synthesis of aminoethanethiol trityl ether ligands for ruthenium-catalysed asymmetric transfer hydrogenation.
Harfouche J, et al.
Tetrahedron Letters, 15(21), 3413-3418 (2004)
Structure and synthesis of a unique isonitrile lipid isolated from the marine mollusk Actinocyclus papillatus.
Manzo E, et al.
Organic Letters, 13(8), 1897-1899 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service