Skip to Content
Merck
All Photos(2)

Key Documents

451142

Sigma-Aldrich

Lithium tetrachloroaluminate

anhydrous, beads, −10 mesh, 99.99% trace metals basis

Synonym(s):

Aluminum lithium chloride (AlLiCl4 ), Lithium aluminum tetrachloride, Lithium tetrachloroaluminate (LiAlCl4 )

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LiAlCl4
CAS Number:
Molecular Weight:
175.73
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

anhydrous

Assay

99.99% trace metals basis

form

beads

reaction suitability

reagent type: catalyst
core: aluminum

particle size

−10 mesh

SMILES string

[Li+].Cl[Al-](Cl)(Cl)Cl

InChI

1S/Al.4ClH.Li/h;4*1H;/q+3;;;;;+1/p-4

InChI key

AQLRWYUVWAYZFO-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Application

A high-energy, new battery system using only inorganic constituents has been designed where Li and LiCoO2 serve as the electrodes in a LiAlCl4-in-SO2 liquid electrolyte. Has been studied for cell performance characteristics and reaction products of LiAlCl4 and LiGaCl4 in a variety of battery systems.
Used in studies of electrochemical cell reactions.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hill, I.R. et al.
Journal of the Electrochemical Society, 142, 3267-3267 (1995)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service