Skip to Content
Merck
All Photos(1)

Key Documents

447161

Sigma-Aldrich

(S)-(−)-3-Benzyloxy-1,2-propanediol

99%

Synonym(s):

(S)-Glycerol 1-benzyl ether, 1-Benzyl-sn-glycerol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2OCH2CH(OH)CH2OH
CAS Number:
Molecular Weight:
182.22
Beilstein:
2415534
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

optical activity

[α]20/D −4.7°, c = 20 in chloroform

bp

261 °C (lit.)

mp

24-26 °C (lit.)

storage temp.

2-8°C

SMILES string

OC[C@H](O)COCc1ccccc1

InChI

1S/C10H14O3/c11-6-10(12)8-13-7-9-4-2-1-3-5-9/h1-5,10-12H,6-8H2/t10-/m0/s1

InChI key

LWCIBYRXSHRIAP-JTQLQIEISA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service