Skip to Content
Merck
All Photos(1)

Documents

446416

Sigma-Aldrich

Diethyl 2-ethyl-2-phenylmalonate

96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C(C2H5)(CO2C2H5)2
CAS Number:
Molecular Weight:
264.32
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

form

liquid

refractive index

n20/D 1.491 (lit.)

bp

185 °C/15 mmHg (lit.)

density

1.07 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)C(CC)(C(=O)OCC)c1ccccc1

InChI

1S/C15H20O4/c1-4-15(13(16)18-5-2,14(17)19-6-3)12-10-8-7-9-11-12/h7-11H,4-6H2,1-3H3

InChI key

PKRVDBARWFJWEB-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Recovery of Arenes Containing Keto or Ester Groups by the Electrochemical Reduction of Their Respective Cyclopentadienyliron Complexes.
Abd-El-Aziz AS, et al.
Synthetic Communications, 19(11-12), 1865-1870 (1989)
The Phenylation of Esters by Reaction with Bromobenzene and Sodium Amide.
Leake WW and Levine R.
Journal of the American Chemical Society, 81(7), 1627-1630 (1959)
STABILITY OF 2-THIOPHENOBARBITAL AND ITS N-METHYL DERIVATIVES IN THE PRESENCE OF ?-CYCLODEXTRIN.
?UCHOWSKI G, et al.
Acta Poloniae Pharmaceutica, 62(5), 335-340 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service