Skip to Content
Merck
All Photos(1)

Key Documents

442348

Sigma-Aldrich

Poly(styrene-co-maleic anhydride), partial isooctyl ester, cumene terminated

average Mn ~2,300 by GPC

Synonym(s):

maleated copolymer, maleated polymer, maleic copolymer

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

form

solid

mol wt

average Mn ~2,300 by GPC

composition

acid groups converted to ester, 50%

acid number

95‑120 mg KOH/g

transition temp

Tg 66 °C

solubility

aqueous base: soluble
ketones and ethers: soluble

General description

~4:1 mole ratio of styrene: maleic anhydride

Application

Leveling aids in films, embrittling/anti-resoil agents in shampoos, pigment dispersants, paper sizing and aqueous grinding fluids.
Poly(styrene-co-maleic anhydride), partial isooctyl ester, cumene terminated (PSMA) may be used in the preparation of inorganic-polymeric nanoparticles (Ag-PSMA) which can be converted to Au based Janus nanoparticles.

Features and Benefits

Low solution viscosity and high functionality.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eccentric inorganic-polymeric nanoparticles formation by thermal induced cross-linked esterification and conversion of eccentricity to raspberry-like Janus.
Li W P, et al.
Chemical Communications (Cambridge, England), 49(16), 1609-1611 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service