Skip to Content
Merck
All Photos(1)

Key Documents

439274

Sigma-Aldrich

2-Chloro-1,3-dimethylimidazolidinium tetrafluoroborate

98%

Synonym(s):

CIB

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H10BClF4N2
CAS Number:
Molecular Weight:
220.40
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:

Assay

98%

mp

175-177 °C (lit.)

SMILES string

F[B-](F)(F)F.CN1CC[N+](C)=C1Cl

InChI

1S/C5H10ClN2.BF4/c1-7-3-4-8(2)5(7)6;2-1(3,4)5/h3-4H2,1-2H3;/q+1;-1

InChI key

UPLXKEIRISBKRM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reagent for:
Esterification and peptide coupling of sterically hindered amino acids
Preparation of efficient diazo-transfer reagents
Oxidative insertion for palladium and nickel catalyst synthesis
Preparation of catalysts for use in coupling reactions
Useful for the esterification and coupling of sterically hindered amino acids.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Proc. 22nd Eur. Pept. Symp.: Peptides 1992, 220-220 (1993)
Proc. 22nd Eur. Pept. Symp.: Peptides 1992, 51-51 (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service