Skip to Content
Merck
All Photos(1)

Key Documents

438235

Sigma-Aldrich

Hydroxylammonium nitrate solution

24 wt. % in H2O, 99.999%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NOH · HNO3
CAS Number:
Molecular Weight:
96.04
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.22

Assay

99.999%

form

liquid

concentration

24 wt. % in H2O

density

1.118 g/mL at 25 °C

SMILES string

NO.O[N+]([O-])=O

InChI

1S/HNO3.H3NO/c2-1(3)4;1-2/h(H,2,3,4);2H,1H2

InChI key

NILJXUMQIIUAFY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 2 Oral

Target Organs

Bone marrow

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service