Skip to Content
Merck
All Photos(1)

Key Documents

437352

Sigma-Aldrich

4-(1-Pyrrolidinyl)piperidine

95%

Synonym(s):

1-(Piperidin-4-yl)pyrrolidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H18N2
CAS Number:
Molecular Weight:
154.25
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

mp

53-56 °C (lit.)

SMILES string

C1CCN(C1)C2CCNCC2

InChI

1S/C9H18N2/c1-2-8-11(7-1)9-3-5-10-6-4-9/h9-10H,1-8H2

InChI key

STWODXDTKGTVCJ-UHFFFAOYSA-N

General description

4-(1-Pyrrolidinyl)piperidine (4-pypp) has been reported to form Hofmann type complexes [M(4-pypp)2Ni(CN)4] (M=Ni or Co) and their FT-IR and Raman spectra have been studied.[1] FT-IR, Raman spectra and the vibrational spectra of 4-pypp have been recorded between 4000 and 400cm-1.[2]

Application

4-(1-Pyrrolidinyl)piperidine was employed for the synthesis of the following analogs:[3]
  • 4-pyrrolidin-1´-yl-1-[2-(2″-nitro-phenyl)-2-oxo-ethyl]-piperidinium bromide
  • 4-pyrrolidin-1´-yl-1-[2-(3″-nitro-phenyl)-2-oxo-ethyl]-piperidinium bromide
  • 4-pyrrolidin-1´-yl-1-[2-(2″,4″-dimethoxy-phenyl)-2-oxo-ethyl]-piperidinium bromide
  • 4-pyrrolidin-1´-yl-1-[2-(3″,4″-dihydroxy-phenyl)-2-oxo-ethyl]-piperidinium bromide
Reactant for synthesis of:
Small molecule ligands of methyl-lysine binding proteins
Small molecules that restore E-cadherin expression and reduce invasion in colorectal carcinoma cells
Selective Polo-like kinase 1 inhibitors
Molecules with effects on plasma glucose levels
Vasopressin1b receptor antagonists
IKKβ inhibitors

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cemal Parlak
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 99, 12-17 (2012-10-09)
Some new Hofmann type complexes with chemical formula of M(4 pypp)(2)Ni(CN)(4) (where 4 py pp=4-(1-pyrrolidinyl)piperidine and MNi or Co) were prepared and their FT-IR and Raman spectra were reported in the region of 4000-200 cm(-1) and 4000-100 cm(-1), respectively. The
Theoretical and experimental vibrational spectroscopic study of 4-(1-Pyrrolidinyl) piperidine.
Parlak C.
Journal of Molecular Structure, 966(1), 1-7 (2010)
Nousheen Mushtaq et al.
Pakistan journal of pharmaceutical sciences, 23(2), 220-223 (2010-04-07)
In the present study some compounds of 4-(1-Pyrrolidinyl) Piperidine (I) have been synthesized. Structures of compounds were confirmed by using HNMR, IR, Mass and UV spectrophotometer techniques. All the derivatives (II, III, IV and V) and the parent compound (I)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service