Skip to Content
Merck
All Photos(3)

Documents

433047

Sigma-Aldrich

4-Methyl-1-indanone

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H10O
CAS Number:
Molecular Weight:
146.19
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

form

solid

mp

94-96 °C (lit.)

SMILES string

Cc1cccc2C(=O)CCc12

InChI

1S/C10H10O/c1-7-3-2-4-9-8(7)5-6-10(9)11/h2-4H,5-6H2,1H3

InChI key

RUORWXQKVXTQJJ-UHFFFAOYSA-N

General description

4-Methyl-1-indanone on condensation with 2-lithio-N,N-diethyl-1-napthamide and 2-lithio-N,N-diethyl-8-methoxy-1-napthamide, followed by hydrolysis afforded 2-(1-hydroxy-4-methylindan-1-yl)-1-naphthoic acid lactone and 2-(1-hydroxy-4-methylindan-1-yl)-8-methoxy-1-naphthoic acid lactone respectively.

Application

4-Methyl-1-indanone may be used for the following syntheses:
  • 3,6-dimethylcholanthrene
  • 3,6-dimethyl-7-methoxycholanthrene
  • 3-methyl-7-methoxycholanthrene
  • methyl 4-methyl-1-fluoroindan-1-carboxylate (4-Me-FICA Me ester)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Syntheses of 3, 6-dimethylcholanthrene, 3, 6-dimethyl-7-methoxycholanthrene and 3-methyl-7-methoxycholanthrene.
Newman MS and Sujeeth PK.
The Journal of Organic Chemistry, 49(16), 2841-2843 (1984)
Jyunichi Koyanagi et al.
Chemical & pharmaceutical bulletin, 62(8), 816-819 (2014-06-13)
An improved synthetic route has been developed for the preparation of methyl 1-fluoroindan-1-carboxylate (FICA Me ester) from 1-indanone. Methyl 4-methyl-1-fluoroindan-1-carboxylate (4-Me-FICA Me ester) was also prepared following the same procedure.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service