Skip to Content
Merck
All Photos(1)

Key Documents

426563

Sigma-Aldrich

1,3-Propanediol bis(4-aminobenzoate)

powder (Granular), 98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2(CH2O2CC6H4NH2)2
CAS Number:
Molecular Weight:
314.34
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

powder (Granular)

eq. wt.

157 Amine

mp

124-127 °C (lit.)

density

1.14 g/mL at 25 °C (lit.)

SMILES string

Nc1ccc(cc1)C(=O)OCCCOC(=O)c2ccc(N)cc2

InChI

1S/C17H18N2O4/c18-14-6-2-12(3-7-14)16(20)22-10-1-11-23-17(21)13-4-8-15(19)9-5-13/h2-9H,1,10-11,18-19H2

InChI key

YPACMOORZSDQDQ-UHFFFAOYSA-N

General description

1,3-Propanediol bis(4-aminobenzoate) (PBA) is an aromatic diamine. Its crystal structure has been determined by single crystal X-ray diffraction. Its ability to improve the flexibility of electrically conductive adhesives (ECAs) has been studied. Results show that it has a good impact on the electrical, mechanical and thermal properties of the ECAs.
Non-hygroscopic, forms urethanes with good hydrolytic stability, dry heat aging and chemical and environmental resistance.

Application

1,3-Propanediol bis(4-aminobenzoate) (PBA) may be used as a flexibilizer, to prepare a flexible epoxy molding compound (EMC) and also as a chain extender in the synthesis of degradable poly(esterurethane urea) (PEUUR).
A safe, high performance reactant with polyurethane and epoxide prepolymers.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adhesion of a novel flexible epoxy molding compound and its molecular dynamics simulation.
Cui HW, et al.
International Journal of Adhesion and Adhesives, 35, 50-54 (2012)
Josh D Erndt-Marino et al.
Journal of biomedical materials research. Part A, 104(9), 2200-2209 (2016-04-22)
This work investigates the potential of cell layer-electrospun mesh constructs as coronary artery bypass grafts. These cell-mesh constructs were generated by first culturing a confluent layer of 10T½ smooth muscle progenitor cells on a high strength electrospun mesh with uniaxially
Satyavrata Samavedi et al.
Acta biomaterialia, 7(12), 4131-4138 (2011-07-28)
Current scaffolds for the regeneration of anterior cruciate ligament injuries are unable to capture intricate mechanical and chemical gradients present in the natural ligament-bone interface. As a result, stress concentrations can develop at the scaffold-bone interface, leading to poor osseointegration.
K Wu et al.
Carcinogenesis, 10(11), 2119-2122 (1989-11-01)
The mutagenic properties of Ethacure 300, Cyanacure and Polacure 740M, all possible substitutes for the industrial carcinogen, 4,4'-methylene bis(2-chloroaniline), have been determined in Salmonella typhimurium strains TA100, TA98, TA1535 and TA1537. These data have been compared with the effects of
Crystal Structure, Optical Properties of Molecular Crystals 1, 3-propanediol Bis (4-aminobenzoate).
Xue RT, et al.
Advanced Materials Research, 1052, 207-211 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service