Skip to Content
Merck
All Photos(1)

Key Documents

42655

Sigma-Aldrich

1,6-Diphenoxy-2,4-hexadiyne

≥98.0%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H14O2
CAS Number:
Molecular Weight:
262.30
Beilstein:
1979094
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0%

form

solid

SMILES string

C(Oc1ccccc1)C#CC#CCOc2ccccc2

InChI

1S/C18H14O2/c1(9-15-19-17-11-5-3-6-12-17)2-10-16-20-18-13-7-4-8-14-18/h3-8,11-14H,15-16H2

InChI key

ZBTRFBNXAKJTBI-UHFFFAOYSA-N

General description

1,6-Diphenoxy-2,4-hexadiyne (dimerized phenyl propargyl ether, DPPE) is a disubstituted diacetylene.[1] It is reported to be synthesized by the oxidative coupling of phenyl propargyl ether.[2] Its structural and spectroscopic properties have been analyzed using vibrational spectroscopy and quantum-chemical calculations.[1] It is reported to form an organic hydrogen getter in combination with 5% Pd-CaCO3.[2] The two crystalline forms of 1,6-diphenoxy-2,4-hexadiyne have been studied.[3]

Application

1,6-Diphenoxy-2,4-hexadiyne may be used to synthesize the trinuclear derivative with an edge bridging ynenyl ligand on reacting with [Ru3(μ-H)(μ3-η2-apyr)(CO)9] (Hapyr=2-aminopyrimidine).[4]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Organic hydrogen getters.
Courtney RL and Harrah LA.
J. Mater. Sci., 12(1), 175-186 (1977)
Two forms of 1,6-diphenoxy-2, 4-hexadiyne and the hydrogenated analog, 1,6-diphenoxyhexane.
Morosin B and Harrah L.
Acta Crystallographica Section B, Structural Crystallography and Crystal Chemistry, 33(6), 1760-1764 (1977)
M M Tauber et al.
Applied and environmental microbiology, 66(4), 1634-1638 (2000-04-01)
Rhodococcus rhodochrous NCIMB 11216 produced nitrile hydratase (320 nkat mg of protein(-1)) and amidase activity (38.4 nkat mg of protein(-1)) when grown on a medium containing propionitrile. These enzymes were able to hydrolyze nitrile groups of both granular polyacrylonitriles (PAN)
η3-Edge-Bridging versus η3-Face-Capping Coordination of a Conjugated Ynenyl Ligand on a Triruthenium Cluster Core.
Cabeza JA, et al.
Organometallics, 23(24), 5849-5855 (2004)
Vibrational and theoretical study of selected diacetylenes.
Roman M and Baranska M.
Spectrochimica Acta. Part A, Molecular and Biomolecular Spectroscopy, 115, 493-503 (2013)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service