Skip to Content
Merck
All Photos(1)

Key Documents

422215

Sigma-Aldrich

2-(Trifluoroacetyl)thiophene

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H3F3OS
CAS Number:
Molecular Weight:
180.15
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

refractive index

n20/D 1.486 (lit.)

bp

163-165 °C (lit.)

density

1.403 g/mL at 25 °C (lit.)

SMILES string

FC(F)(F)C(=O)c1cccs1

InChI

1S/C6H3F3OS/c7-6(8,9)5(10)4-2-1-3-11-4/h1-3H

InChI key

CZYKJGCKVBXLGF-UHFFFAOYSA-N

General description

2-(Trifluoroacetyl)thiophene is a heterocyclic trifluoromethyl ketone. Its asymmetric reduction to alcohols with moderate enantioselectivities catalyzed by Arabidopsis thaliana seedlings has been reported.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

138.2 °F - closed cup

Flash Point(C)

59 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Arabidopsis thaliana: a novel biocatalyst for asymmetric reductions.
Plant Biotechnology, 28(1), 77-82 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service