Skip to Content
Merck
All Photos(1)

Key Documents

413313

Sigma-Aldrich

Tris(4-hydroxyphenyl)methane triglycidyl ether 2,6-tolylene diisocyanate adduct

Synonym(s):

Triphenylolmethane triglycidyl ether 2,6-tolylene diisocyanate adduct

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C65H62N2O14
CAS Number:
Molecular Weight:
1095.19
UNSPSC Code:
12162002
PubChem Substance ID:

mp

90 °C (lit.)

SMILES string

Cc1c(cccc1N2CC(COc3ccc(cc3)C(c4ccc(OCC5CO5)cc4)c6ccc(OCC7CO7)cc6)OC2=O)N8CC(COc9ccc(cc9)C(c%10ccc(OCC%11CO%11)cc%10)c%12ccc(OCC%13CO%13)cc%12)OC8=O

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service