Skip to Content
Merck
All Photos(1)

Documents

409863

Sigma-Aldrich

Bisphenol A propoxylate

average Mn ~344, PO/phenol 1

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C[C6H4[OCH2CH(CH3)]nOH]2
CAS Number:
UNSPSC Code:
12162002
PubChem Substance ID:

mol wt

average Mn ~344

composition

PO/phenol, 1

refractive index

n20/D 1.555 (lit.)

density

1.09 g/mL at 25 °C (lit.)

SMILES string

OCCCO.CC(C)(c1ccc(O)cc1)c2ccc(O)cc2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service