Skip to Content
Merck
All Photos(1)

Key Documents

407313

Sigma-Aldrich

Methyl trans-3-pentenoate

90%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH=CHCH2CO2CH3
CAS Number:
Molecular Weight:
114.14
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

90%

form

liquid

refractive index

n20/D 1.421 (lit.)

bp

55-56 °C/20 mmHg (lit.)

density

0.93 g/mL at 25 °C (lit.)

SMILES string

[H]\C(C)=C(\[H])CC(=O)OC

InChI

1S/C6H10O2/c1-3-4-5-6(7)8-2/h3-4H,5H2,1-2H3/b4-3+

InChI key

KJALUUCEMMPKAC-ONEGZZNKSA-N

General description

Methyl trans-3-pentenoate is an ester. Ring-closing metathesis (RCM) of methyl trans-3-pentenoate using silica- and monolith supported Grubbs-Herrmann-type catalysts has been reported. The 1,3-dipolar cycloaddition of with C-ethoxycarbonyl nitrone to form isoxazolidines has been studied.

Application

Methyl trans-3-pentenoate may be used for the total synthesis of phytochemicals (-)-grandinolide and (-)-sapranthin.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

75.2 °F - closed cup

Flash Point(C)

24 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Total Syntheses of (-)-Grandinolide and (-)-Sapranthin by the Sharpless Asymmetric Dihydroxylation of Methyl trans-3-Pentenoate: Elucidation of the Stereostructure of (-)-Sapranthin.
Harcken C, et al.
Chemistry (Weinheim An Der Bergstrasse, Germany), 4(11), 2342-2352 (1998)
Monolith-and Silica-Supported Carboxylate-Based Grubbs-Herrmann-Type Metathesis Catalysts.
Krause JO, et al.
Advanced Synthesis & Catalysis, 345(8), 996-1004 (2003)
C-Alkoxycarbonyl nitrones: building blocks for the synthesis of butenolides, lactams and modified nucleosides.
Romeo G, et al.
Mini-Reviews in Organic Chemistry, 2(1), 59-77 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service