Skip to Content
Merck
All Photos(1)

Key Documents

404853

Sigma-Aldrich

cis-4-Aminocyclohexanecarboxylic acid

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6H10CO2H
CAS Number:
Molecular Weight:
143.18
Beilstein:
2802522
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

95%

form

powder

mp

299-301 °C (lit.)

SMILES string

N[C@H]1CC[C@H](CC1)C(O)=O

InChI

1S/C7H13NO2/c8-6-3-1-5(2-4-6)7(9)10/h5-6H,1-4,8H2,(H,9,10)/t5-,6+

InChI key

DRNGLYHKYPNTEA-OLQVQODUSA-N

Looking for similar products? Visit Product Comparison Guide

General description

cis-4-aminocyclohexanecarboxylic acid (C4-ACHC, cis-ACCA) exists in zwitterionic form. Cyclohexane ring in the molecule of cis-4-aminocyclohexanecarboxylic acid is present in chair conformation. The carboxylate and ammonium groups in the molecule occupy axial and equatorial positions, respectively.[1] It co-crystallizes with water molecules in a 2:1 (amino acid:water ratio).[1] H-Cys-Leu-Gly-Gly-Leu-Leu-Thr-Met-Val-OH (CLG) peptide analogue containing cis-ACCA has been prepared and its activity has been studied.[2]

Application

cis-4-Aminocyclohexanecarboxylic acid may be used in the synthesis of new analogs of arginine vasopressin (AVP).[3] It may be used in the synthesis of cis-4-[[[(2-chloroethyl)nitrosoamino]carbonyl]methylamino]cyclohexanecarboxylic acid.[4]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hydrogen-bonding patterns in cis-4-ammoniocyclohexanecarboxylate hemihydrate.
Edward E Avila et al.
Acta crystallographica. Section C, Crystal structure communications, 60(Pt 10), o759-o761 (2004-10-07)
T P Johnston et al.
Journal of medicinal chemistry, 27(1), 97-99 (1984-01-01)
cis-4-[[[(2-Chloroethyl)nitrosoamino]carbonyl]methylamino] cyclohexanecarboxylic acid (N-Me-cis-CCCNU) was synthesized in five steps from cis-4-aminocyclohexanecarboxylic acid via an N-tosylated intermediate. N-Me-cis-CCCNU, which is incapable of the facile decomposition that characterizes the clinically useful nitrosoureas, effected a significant cure rate of both early and established
S K Lo et al.
Journal of applied physiology (Bethesda, Md. : 1985), 58(5), 1421-1427 (1985-05-01)
We examined the effect of fibrinolysis depression on thrombin-induced pulmonary microembolism in awake sheep prepared with chronic lung lymph fistulas. Fibrinolysis was depressed by an intravenous infusion (100 mg) of tranexamic acid [trans-4-(Aminomethyl)cyclohexanecarboxylic acid]. Pulmonary microembolism was induced by an
Dariusz Sobolewski et al.
Protein and peptide letters, 14(3), 213-217 (2007-03-10)
Synthesis of thirteen new analogues of arginine vasopressin (AVP) has been described. Amino acid residues at positions 2 and 3 of AVP, [3-mercaptopropionic acid (Mpa)(1)]AVP (dAVP), [Mpa(1),d-Arg(8)]VP (dDAVP) and [Mpa(1),Val(4),d-Arg(8)]VP (dVDAVP) were replaced with one amino acid residue using sterically
Mauro Marastoni et al.
Bioorganic & medicinal chemistry, 10(9), 3061-3066 (2002-07-12)
In order to improve the immunotherapeutical potential of H-Cys-Leu-Gly-Gly-Leu-Leu-Thr-Met-Val-OH (CLG) peptide, an Epstein-Barr virus (EBV) subdominant epitope derived from the membrane protein LMP2, we have synthesized and tested CLG analogues containing cis- and/or trans-4-aminocyclohexanecarboxylic acid (ACCA) replacing Gly-Gly and/or Thr-Met

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service