Skip to Content
Merck
All Photos(1)

Documents

400106

Sigma-Aldrich

Thallium(III) acetate sesquihydrate

Synonym(s):

Thallic acetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3COO)3Tl · 1.5H2O
CAS Number:
Molecular Weight:
291.46
Beilstein:
3693590
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

form

solid

mp

182 °C (dec.) (lit.)

SMILES string

O.CC(=O)O[Tl](OC(C)=O)OC(C)=O

Looking for similar products? Visit Product Comparison Guide

Other Notes

Hydration may vary slightly

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service