379085
Diethyl 5,5′-methylenebis(4-ethyl-3-methyl-2-pyrrolecarboxylate)
97%
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C21H30N2O4
CAS Number:
Molecular Weight:
374.47
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay
97%
mp
128-129 °C (lit.)
functional group
ester
SMILES string
CCOC(=O)c1[nH]c(Cc2[nH]c(C(=O)OCC)c(C)c2CC)c(CC)c1C
InChI
1S/C21H30N2O4/c1-7-14-12(5)18(20(24)26-9-3)22-16(14)11-17-15(8-2)13(6)19(23-17)21(25)27-10-4/h22-23H,7-11H2,1-6H3
InChI key
BPMQEODKNZPRMH-UHFFFAOYSA-N
General description
Diethyl 5,5′-methylenebis(4-ethyl-3-methyl-2-pyrrolecarboxylate) is a heterocyclic building block.
Application
Diethyl 5,5′-methylenebis(4-ethyl-3-methyl-2-pyrrolecarboxylate) may be used as starting reagent for the synthesis of bis-pyrrolylmethane based anion receptors.
Diethyl 5,5′-methylenebis(4-ethyl-3-methyl-2-pyrrolecarboxylate) may be used in the preparation of 2,2′-bisamidodipyrrolylmethanes.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Anion complexation properties of 2, 2'-bisamidodipyrrolylmethanes.
El DrubiaVega I.
Chemical Communications (Cambridge, England), 14, 1686-1687 (2003)
Gloe K.
Macrocyclic Chemistry: Current Trends and Future Perspectives., 162-162 (2005)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service