Skip to Content
Merck
All Photos(1)

Key Documents

374717

Sigma-Aldrich

Triethyl 2-fluoro-2-phosphonoacetate

96%

Synonym(s):

2-Fluoro-2-phosphonoacetic acid triethyl ester, Diethyl [(ethoxycarbonyl)fluoromethyl]phosphonate, Ethyl diethylphosphonofluoroacetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5O)2P(O)CH(F)CO2C2H5
CAS Number:
Molecular Weight:
242.18
Beilstein:
1912161
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

form

liquid

reaction suitability

reaction type: C-C Bond Formation

refractive index

n20/D 1.425 (lit.)

bp

75 °C/0.01 mmHg (lit.)

density

1.194 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)C(F)P(=O)(OCC)OCC

InChI

1S/C8H16FO5P/c1-4-12-8(10)7(9)15(11,13-5-2)14-6-3/h7H,4-6H2,1-3H3

InChI key

FVPISMANESAJQZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for:
  • Diels-Alder reactions
  • Biosynthesis of terpene
  • Stereoselective synthesis of unsaturated esters, fluorides and nitriles from the reactions of aldehydes and ketones using Wadsworth-Emmons phosphonates
  • Synthesis of quinolones
  • Horner-Wadsworth-Emmons asymmetric hydration
  • Addition reactions and the subsequent application to click chemistry
  • Asymmetric intermolecular Stetter reactions

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

163.4 °F - closed cup

Flash Point(C)

73 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P W Collins et al.
Journal of medicinal chemistry, 30(11), 1952-1955 (1987-11-01)
A 4-fluoro analogue of enisoprost was prepared and evaluated for gastric antisecretory and mucosal protective activity in animals. The synthesis centered upon cuprate chemistry but also involved a Wittig reaction to produce a cis fluoro olefinic moiety, a furan rearrangement/isomerization

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service