Skip to Content
Merck
All Photos(1)

Key Documents

36615

Sigma-Aldrich

Dicyanocobyrinic acid heptamethyl ester

≥95.0%

Synonym(s):

Heptamethyl dicyano cobyrinate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C54H73CoN6O14
CAS Number:
Molecular Weight:
1089.12
Beilstein:
1207601
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95.0%

mp

124-125 °C (dec.)

storage temp.

2-8°C

SMILES string

[H][C@]12[C@H](CC(=O)OC)[C@@](C)(CCC(=O)OC)C(N1[Co](C#N)C#N)=C(C)C3=NC(=CC4=NC(=C(C)C5=N[C@]2(C)[C@@](C)(CC(=O)OC)[C@@H]5CCC(=O)OC)[C@@](C)(CC(=O)OC)[C@@H]4CCC(=O)OC)C(C)(C)[C@@H]3CCC(=O)OC

InChI

1S/C52H73N4O14.2CN.Co/c1-28-43-31(17-20-37(58)65-10)48(3,4)35(54-43)25-34-30(16-19-36(57)64-9)50(6,26-41(62)69-14)46(53-34)29(2)44-32(18-21-38(59)66-11)51(7,27-42(63)70-15)52(8,56-44)47-33(24-40(61)68-13)49(5,45(28)55-47)23-22-39(60)67-12;2*1-2;/h25,30-33,47H,16-24,26-27H2,1-15H3;;;/q-1;;;+1/t30-,31-,32-,33+,47-,49-,50+,51+,52+;;;/m1.../s1

InChI key

CPBDBBXBECBXRW-NTQATXMTSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Preparation of chloro derivatives of dicyanocobyrinic acid heptamethyl ester (′cobester′) is reported.[1]

Application

Dicyanocobyrinic acid heptamethyl ester may be used in the preparation of (5R,6R)-Coa,Coβ-dicyano-5,6-dihydro-5-hydroxy-heptamethylcob(III)yrinate-c,6-lactone, a derivative of cobyrinic acid heptamethyl ester.[2]

Other Notes

Stable derivative of cobyrinic acid; used e.g. as catalyst for carbon-skeleton rearrangements [3]; Partial ester hydrolysis and prepn. of derivatives[4][5][6]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y. Murakami et al.
Chemistry Letters (Jpn), 469-469 (1988)
H H Inhoffen et al.
Philosophical transactions of the Royal Society of London. Series B, Biological sciences, 273(924), 327-333 (1976-02-05)
Preparations of C10 acetate, trifluoroacetate, and chloro derivatives of dicyanocobyrinic acid heptamethyl ester ('cobester') are described. The nature and properties of intermediates in the synthesis of the 10-chloro derivative are discussed, and the electronic absorption spectra of the products are
Susan M Chemaly et al.
Inorganic chemistry, 50(18), 8700-8718 (2011-08-20)
A stable yellow derivative of cobyrinic acid heptamethyl ester, (5R,6R)-Coα,Coβ-dicyano-5,6-dihydro-5-hydroxy-heptamethylcob(III)yrinate-c,6-lactone (DCSYCbs), was prepared from dicyanocobyrinic acid heptamethyl ester (DCCbs). The C5 carbon is oxidized and the c side chain cyclized to form a lactone at C6; the 13 atom, 14
B. Gruning et al.
Helvetica Chimica Acta, 68, 1771-1771 (1985)
Y. Murakami et al.
Chemistry Letters (Jpn), 477-477 (1985)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service