Skip to Content
Merck
All Photos(1)

Documents

365106

Sigma-Aldrich

Bismuth strontium calcium copper oxide

powder, 5 μm

Synonym(s):

BSCCO 2212

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Bi2Sr2CaCu2O8+X(x=0.15-0.20)
CAS Number:
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:

form

powder

particle size

5 μm

density

6.4 g/mL at 25 °C (lit.)

SMILES string

O=[Ca].O=[Cu].O=[Cu].O=[Sr].O=[Sr].O=[Bi]O[Bi]=O.O=[Bi]O[Bi]=O

InChI

1S/4Bi.Ca.2Cu.11O.2Sr

InChI key

SCYGCYNQMSDOOE-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service