Skip to Content
Merck
All Photos(1)

Key Documents

363200

Sigma-Aldrich

4-(Octyloxy)benzoic acid

98%

Synonym(s):

4-n -Octyloxybenzoic acid, p -Octoxybenzoic acid, p -(Octyloxy)benzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)7OC6H4CO2H
CAS Number:
Molecular Weight:
250.33
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

98%

form

liquid crystal

mp

101-105 °C (lit.)

transition temp

crystalline phase to smectic phase 101 °C
smectic phase to nematic phase 108 °C
nematic phase to isotropic phase 147 °C

SMILES string

CCCCCCCCOc1ccc(cc1)C(O)=O

InChI

1S/C15H22O3/c1-2-3-4-5-6-7-12-18-14-10-8-13(9-11-14)15(16)17/h8-11H,2-7,12H2,1H3,(H,16,17)

InChI key

IALWCYFULVHLEC-UHFFFAOYSA-N

Application

Intermediate for the synthesis of liquid crystals including chiral ferroelectric benzoates. Used in the preparation of medicinal compounds.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aldrichimica Acta, 23, 23-23 (1990)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service