Skip to Content
Merck
All Photos(1)

Key Documents

349747

Sigma-Aldrich

Ethyl (2-methylphenoxy)acetate

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4OCH2CO2C2H5
CAS Number:
Molecular Weight:
194.23
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

refractive index

n20/D 1.503 (lit.)

bp

119-120 °C (lit.)

density

1.073 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)COc1ccccc1C

InChI

1S/C11H14O3/c1-3-13-11(12)8-14-10-7-5-4-6-9(10)2/h4-7H,3,8H2,1-2H3

InChI key

NHFQSFIBSJPLIW-UHFFFAOYSA-N

General description

Ethyl (2-methylphenoxy)acetate is an ester.

Application

Ethyl (2-methylphenoxy)acetate may be used in chemical synthesis.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service