Skip to Content
Merck
All Photos(1)

Key Documents

345717

Sigma-Aldrich

1-Bromo-2,3-difluorobenzene

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H3F2
CAS Number:
Molecular Weight:
192.99
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

form

liquid

refractive index

n20/D 1.509 (lit.)

density

1.724 g/mL at 25 °C (lit.)

SMILES string

Fc1cccc(Br)c1F

InChI

1S/C6H3BrF2/c7-4-2-1-3-5(8)6(4)9/h1-3H

InChI key

RKWWASUTWAFKHA-UHFFFAOYSA-N

General description

Microbial oxidation of 1-bromo-2,3-difluorobenzene by Pseudomonas putida strain 39/D and Escherichia coli recombinant microorganism (strain JM 109(pDTG601)) has been reported.[1]

Application

1-Bromo-2,3-difluorobenzene may be used in the synthesis of potent, orally active Calcitonin gene-related peptide (CGRP) receptor antagonist (BMS-846372).[2] It may be used in the preparation of 2,3-difluorophenyl(dimethyl)phosphane.[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

141.8 °F - closed cup

Flash Point(C)

61 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reaction of chloropentafluorobenzene and 2, 3-and 2, 6-difluorobromobenzenes with lithium dimethylphosphide.
Goryunov LI, et al.
Russ. J. Org. Chem., 45(12), 1859-1861 (2009)
New metabolites from the microbial oxidation of fluorinated aromatic compounds.
Hudlicky T, et al.
Journal of Fluorine Chemistry, 89(1), 23-30 (1998)
Guanglin Luo et al.
ACS medicinal chemistry letters, 3(4), 337-341 (2012-04-12)
Calcitonin gene-related peptide (CGRP) receptor antagonists have been clinically shown to be effective in the treatment of migraine, but identification of potent and orally bioavailable compounds has been challenging. Herein, we describe the conceptualization, synthesis, and preclinical characterization of a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service