Skip to Content
Merck
All Photos(1)

Key Documents

338419

Sigma-Aldrich

(R)-(+)-3-Chloro-1-phenyl-1-propanol

98%

Synonym(s):

α-(2-Chloroethyl)benzyl alcohol, (R)-(+)-α-(2-Chloroethyl)benzyl alcohol, (R)-(+)-3-Chloro-1-phenylpropanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2CH2CH(C6H5)OH
CAS Number:
Molecular Weight:
170.64
Beilstein:
5250766
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

optical activity

[α]24/D +26°, c = 1 in chloroform

optical purity

ee: 99% (GLC)

mp

58-60 °C (lit.)

SMILES string

O[C@H](CCCl)c1ccccc1

InChI

1S/C9H11ClO/c10-7-6-9(11)8-4-2-1-3-5-8/h1-5,9,11H,6-7H2/t9-/m1/s1

InChI key

JZFUHAGLMZWKTF-SECBINFHSA-N

Application

(R)-(+)-3-Chloro-1-phenyl-1-propanol may be used as a building block in the synthesis of antidepressants such as (R)- and (S)-tomoxetine, fluoxetine and nisoxetine.[1] It may also be used in the synthesis of biologically active 2-substituted chromans such as tephrowatsin E.[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An efficient route to enantiomerically pure antidepressants: tomoxetine, nisoxetine and fluoxetine.
Schneider MP and Goergens U.
Tetrahedron Asymmetry, 3(4), 525-528 (1992)
A stereocontrolled route to 2-substituted chromans.
Hodgetts KJ.
Tetrahedron Letters, 41(44), 8655-8659 (2000)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service