Skip to Content
Merck
All Photos(1)

Documents

335088

Sigma-Aldrich

Hydridotetrakis(triphenylphosphine)rhodium(I)

Synonym(s):

HRh(PPh3)4, Tetrakis(triphenylphosphine)rhodium hydride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(C6H5)3P]4RhH
CAS Number:
Molecular Weight:
1153.06
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

reaction suitability

reagent type: catalyst
core: rhodium

mp

140-146 °C (lit.)

SMILES string

[RhH].c1ccc(cc1)P(c2ccccc2)c3ccccc3.c4ccc(cc4)P(c5ccccc5)c6ccccc6.c7ccc(cc7)P(c8ccccc8)c9ccccc9.c%10ccc(cc%10)P(c%11ccccc%11)c%12ccccc%12

InChI

1S/4C18H15P.Rh.H/c4*1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18;;/h4*1-15H;;

InChI key

QTTJMGBPWXLZKV-UHFFFAOYSA-N

Application

Catalyst for hydrogenation of alkenes and hydrosilylation of alkynes.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Beaupere, D.; Bauer, P.; Uzan, R.
Canadian Journal of Chemistry, 57, 218-218 (1979)
Watanabe, H.; Asami, M.; Nagai, Y.
Journal of Organometallic Chemistry, 195, 363-363 (1980)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service