Skip to Content
Merck
All Photos(1)

Key Documents

333891

Sigma-Aldrich

Silane

≥99.998%, electronic grade

Synonym(s):

Silicon tetrahydride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SiH4
CAS Number:
Molecular Weight:
32.12
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

grade

electronic grade

vapor density

1.1 (vs air)

Assay

≥99.998%

form

gas

resistivity

>1000 Ω-cm

impurities

<1 ppm Carbon dioxide (CO2)
<1 ppm Carbon monoxide (CO)
<1 ppm Chlorosilanes
<1 ppm THC
<1 ppm Trisilane (Si3H8)
<1 ppm Water (H2O)
<100 ppm Hydrogen (H2)
<2 ppm Nitrogen (N2)
<2 ppm Siloxanes
<5 ppm Disilane (Si2H6)

bp

−112 °C (lit.)

mp

−185 °C (lit.)

transition temp

critical temperature −3.4 °C

density

1.114 g/mL at 25 °C (lit.)

SMILES string

[SiH4]

Looking for similar products? Visit Product Comparison Guide

Application

Silane is used in the deposition of amorphous silicon, epitaxial silicon and silicon based dielectrics. Polycrystalline films can be deposited on silicon wafers and glass substrates via remote plasma chemical vapor deposition (RPCVD) using a SiH4-SiF2-H2 gas mixture. Silane is used widely as a dopant in the formation of III-IV semiconductor materials.

Recommended products

Stainless steel regulators Z527416 or Z527424 are recommended.

Pictograms

FlameGas cylinder

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Gas 1A - Press. Gas Compr. Gas

Storage Class Code

2A - Gases

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hu, C. C.
Thin Solid Films, 288, 147-147 (1996)
Quinn, L.J.
Thin Solid Films, 296, 7-7 (1997)
Structural and electrical properties of low temperature polycrystalline silicon deposited using SiF4?SiH4?H2
Jin Jang
Thin Solid Films, 289, 227-227 (1996)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service