Skip to Content
Merck
All Photos(1)

Documents

316369

Sigma-Aldrich

Diethyl(3-pyridyl)borane

97%

Synonym(s):

3-(Diethylboryl)pyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H14BN
CAS Number:
Molecular Weight:
147.03
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

reaction suitability

reagent type: reductant

mp

172-175 °C (lit.)

SMILES string

CCB(CC)c1cccnc1

InChI

1S/C9H14BN/c1-3-10(4-2)9-6-5-7-11-8-9/h5-8H,3-4H2,1-2H3

InChI key

OJKBCQOJVMAHDX-UHFFFAOYSA-N

Application

Reactant for:
  • Preparation of inhibitors of ataxia telangiectasia mutated and Rad3 related (ATR) protein kinase as potential anticancer agents
  • N-Methylation reactions of the pyridyl group
  • Palladium catalyzed amination reactions
  • Intramolecular Heck reactions
  • Coupling reactions
  • Synthesis of trisubstituted pyrimidines from polyhalopyrimidines via Suzuki coupling

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service