Skip to Content
Merck
All Photos(1)

Documents

308226

Sigma-Aldrich

Ethyl isocyanatoformate

90%, technical grade

Synonym(s):

Ethoxycarbonyl isocyanate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
OCNCO2C2H5
CAS Number:
Molecular Weight:
115.09
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Assay

90%

form

liquid

refractive index

n20/D 1.408 (lit.)

bp

25 °C/10 mmHg (lit.)

density

1.115 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CCOC(=O)N=C=O

InChI

1S/C4H5NO3/c1-2-8-4(7)5-3-6/h2H2,1H3

InChI key

VMVZGGPZNHFGKS-UHFFFAOYSA-N

Related Categories

Application

Ethyl isocyanatoformate was used in the synthesis of ethyl[(phenylamino)carbonyl]carbamate by reacting with aniline.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

96.8 °F - closed cup

Flash Point(C)

36 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lin Zhou et al.
Chemical research in toxicology, 17(3), 416-423 (2004-03-18)
Whereas nitrosation of secondary amines produces nitrosamines, amino acids with primary amino groups and glycine ethyl ester were reported to react with nitrite to give unidentified agents that alkylated 4-(p-nitrobenzyl)pyridine to produce purple dyes and be direct mutagens in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service