Skip to Content
Merck
All Photos(1)

Key Documents

307882

Sigma-Aldrich

(R)-(−)-2-Phenylglycine methyl ester hydrochloride

≥95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH(NH2)CO2CH3·HCl
CAS Number:
Molecular Weight:
201.65
EC Number:
MDL number:
UNSPSC Code:
12352200
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95%

form

solid

optical activity

[α]20/D −118°, c = 1 in H2O

reaction suitability

reaction type: solution phase peptide synthesis

mp

189-191 °C (lit.)

application(s)

peptide synthesis

SMILES string

Cl.COC(=O)[C@H](N)c1ccccc1

InChI

1S/C9H11NO2.ClH/c1-12-9(11)8(10)7-5-3-2-4-6-7;/h2-6,8H,10H2,1H3;1H/t8-;/m1./s1

InChI key

DTHMTBUWTGVEFG-DDWIOCJRSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Ivan Muñoz et al.
Water research, 40(19), 3533-3540 (2006-09-23)
A comparative life cycle assessment (LCA) of two solar-driven advanced oxidation processes, namely heterogeneous semiconductor photocatalysis and homogeneous photo-Fenton, both coupled to biological treatment, is carried out in order to identify the environmentally preferable alternative to treat industrial wastewaters containing
Wei Du et al.
Biotechnology and applied biochemistry, 38(Pt 2), 107-110 (2003-05-17)
Ammonium, provided by ammonium carbamate as a novel acyl acceptor, was adopted for enzymic enantioselective ammonolysis of racemic phenylglycine methyl ester in this paper and it has been found that the reaction conditions have profound effects on enzymic activity and
R Fernández-Lafuente et al.
Biomacromolecules, 2(1), 95-104 (2001-12-26)
The importance of the stabilization of the quaternary structure of multimeric enzymes has been illustrated using a model reaction with great industrial relevance: the enzymatic synthesis of ampicillin from 6-amino penicillanic acid (6APA) and phenylglycine methyl ester (PGM) catalyzed by
Wei Du et al.
Sheng wu gong cheng xue bao = Chinese journal of biotechnology, 18(2), 242-245 (2002-08-01)
A novel reaction-enzymatic ammonolysis discovered in the mid of 1990s has been demonstrated to be a very promising alternative in the preparation of optically pure compounds. The effects of organic solvent, initial water activity, temperature and additives on lipase Novozym435-catalyzed
Alessandro Moretto et al.
Chirality, 17(8), 481-487 (2005-08-23)
Reactions of a racemic amine with chiral, N(alpha)-acetylated, C(alpha)-methyl l-phenylglycine-based dipeptide 5(4H)-oxazolones proceed diastereoselectively to give predominantly dipeptide alkylamides comprising d-alpha-phenylethylamine. Diastereoselectivity is remarkably sensitive to solvent polarity and reaction temperature but not significantly to the nature of the C(alpha)-tetrasubstituted

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service