307262
2,4-Dinitrobenzyl chloride
99%
Synonym(s):
α-Chloro-2,4-dinitrotoluene
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
(O2N)2C6H3CH2Cl
CAS Number:
Molecular Weight:
216.58
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
99%
mp
34-36 °C (lit.)
SMILES string
[O-][N+](=O)c1ccc(CCl)c(c1)[N+]([O-])=O
InChI
1S/C7H5ClN2O4/c8-4-5-1-2-6(9(11)12)3-7(5)10(13)14/h1-3H,4H2
InChI key
DARDYTBLZQDXBK-UHFFFAOYSA-N
Application
2,4-Dinitrobenzyl chloride was used in the synthesis of 2-(2,4-dinitrobenzylthio) and 2-(2,4-dinitrobenzylseleno) derivatives of 4,5-dihydroimidazolium chloride.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Corr. 1B
Storage Class Code
8B - Non-combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
235.4 °F - closed cup
Flash Point(C)
113 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Synthesis of 6aλ4?thia?1, 6-diselena-3, 4-diazapentalenes, 1, 6, 6aλ4-triselena-3, 4-diazapentalenes, 6aλ4-thia-1, 3, 4, 6-tetraazapentalenes, and 6aλ4-Selena-1, 3, 4, 6-tetraazapentalenes from cyclic thioureas and selenoureas.
Lai LL and Reid DH.
Heteroatom Chem., 8(1), 13-27 (1997)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service