Skip to Content
Merck
All Photos(1)

Key Documents

305340

Sigma-Aldrich

Copper(II) nitrate on Celite®

extent of labeling: 30 wt. % loading

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cu(NO3)2
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
powder and chunks

grade

for analytical purposes

form

powder and chunks

concentration

28.0-34.0% (with Na2S2O3, titration)

extent of labeling

30 wt. % loading

application(s)

battery manufacturing

SMILES string

[Cu++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/Cu.2NO3/c;2*2-1(3)4/q+2;2*-1

InChI key

XTVVROIMIGLXTD-UHFFFAOYSA-N

Legal Information

Celite is a registered trademark of Imerys Minerals California, Inc.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Met. Corr. 1 - Ox. Sol. 1 - Skin Corr. 1B - STOT RE 1 Inhalation

Target Organs

Lungs

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service