Skip to Content
Merck
All Photos(1)

Key Documents

298573

Sigma-Aldrich

4-Dimethylamino-2,2,6,6-tetramethylpiperidine

96%

Synonym(s):

N,N,2,2,6,6-Hexamethyl-4-piperidinamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H24N2
CAS Number:
Molecular Weight:
184.32
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

refractive index

n20/D 1.4641 (lit.)

SMILES string

CN(C)C1CC(C)(C)NC(C)(C)C1

InChI

1S/C11H24N2/c1-10(2)7-9(13(5)6)8-11(3,4)12-10/h9,12H,7-8H2,1-6H3

InChI key

LRAJIZNKBMCWJE-UHFFFAOYSA-N

Gene Information

Application

4-Dimethylamino-2,2,6,6-tetramethylpiperidine may be used in chemical synthesis.
Reactant for:
Self-aggregation of supramolecules of nitroxides
Synthesis of unsymmetrical spin-labeled bolaamphiphiles

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

120.2 °F - closed cup

Flash Point(C)

49 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service