Skip to Content
Merck
All Photos(1)

Key Documents

294195

Sigma-Aldrich

(1S,2S)-(+)-Pseudoephedrine sulfate salt

98%

Synonym(s):

α-(1-Methylaminoethyl)benzyl alcohol sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H32N2O6S
CAS Number:
Molecular Weight:
428.54
EC Number:
MDL number:
Pricing and availability is not currently available.

Assay

98%

optical activity

[α]25/D +48°, c = 0.8 in ethanol

mp

175-179 °C (lit.)

SMILES string

OS(O)(=O)=O.CN[C@@H](C)[C@@H](O)c1ccccc1.CN[C@@H](C)[C@@H](O)c2ccccc2

InChI

1S/2C10H15NO.H2O4S/c2*1-8(11-2)10(12)9-6-4-3-5-7-9;1-5(2,3)4/h2*3-8,10-12H,1-2H3;(H2,1,2,3,4)/t2*8-,10+;/m00./s1

InChI key

CAVQBDOACNULDN-NRCOEFLKSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service