Skip to Content
Merck
All Photos(1)

Documents

29347

Sigma-Aldrich

N-Cyclohexylaniline

≥98.0% (GC/NT)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H17N
CAS Number:
Molecular Weight:
175.27
Beilstein:
2209864
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0% (GC/NT)

form

liquid

refractive index

n20/D 1.560

density

0.996 g/mL at 20 °C (lit.)

SMILES string

C1CCC(CC1)Nc2ccccc2

InChI

1S/C12H17N/c1-3-7-11(8-4-1)13-12-9-5-2-6-10-12/h1,3-4,7-8,12-13H,2,5-6,9-10H2

InChI key

TXTHKGMZDDTZFD-UHFFFAOYSA-N

Application

N-Cyclohexylaniline has been used as reagent in palladium-catalyzed amination of aromatic bromides.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 4 Oral - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

276.8 °F - closed cup

Flash Point(C)

136 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mahavir Prashad et al.
The Journal of organic chemistry, 68(3), 1163-1164 (2003-02-01)
An efficient palladium-catalyzed amination of aromatic bromides with hindered N-alkyl-substituted anilines is described, either using the combination of Pd(OAc)(2) and P(t-Bu)(3) or a palladium(I) tri-tert-butylphosphine bromide dimer, [Pd(mu-Br)(t-Bu(3)P)](2), a new, commercially available, and easily handled catalyst.
Edmar Ramos Oliveira-Filho et al.
Journal of molecular microbiology and biotechnology, 28(5), 225-235 (2019-02-21)
Three different polyhydroxyalkanoate (PHA) synthase genes (Ralstonia eutropha H16, Aeromonas sp. TSM81 or Aeromonas hydrophila ATCC7966 phaC) were introduced into the chromosome of two Pseudomonas strains: a native medium-chain-length 3-polyhydroxyalkanoate (PHAMCL) producer (Pseudomonas sp. LFM046) and a UV-induced mutant strain

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service