Skip to Content
Merck
All Photos(1)

Key Documents

28556

Sigma-Aldrich

Dichloro(3-cyanopropyl)methylsilane

≥97.0% (GC)

Synonym(s):

(3-Cyanopropyl)dichloro-methylsilane, (3-Cyanopropyl)methyl-dichlorosilane, 4-(Methyldichlorosilyl)butyronitrile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H9Cl2NSi
CAS Number:
Molecular Weight:
182.12
Beilstein:
1750770
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:

Assay

≥97.0% (GC)

refractive index

n20/D 1.458

bp

79 °C/1 mmHg (lit.)

density

1.150 g/mL at 20 °C (lit.)

SMILES string

C[Si](Cl)(Cl)CCCC#N

InChI

1S/C5H9Cl2NSi/c1-9(6,7)5-3-2-4-8/h2-3,5H2,1H3

InChI key

UIFBMBZYGZSWQE-UHFFFAOYSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

197.6 °F - closed cup

Flash Point(C)

92 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service