Skip to Content
Merck
All Photos(1)

Documents

284629

Sigma-Aldrich

Amitriptyline hydrochloride

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H23N · HCl
CAS Number:
Molecular Weight:
313.86
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Assay

99%

SMILES string

Cl[H].CN(C)CC\C=C1\c2ccccc2CCc3ccccc13

InChI

1S/C20H23N.ClH/c1-21(2)15-7-12-20-18-10-5-3-8-16(18)13-14-17-9-4-6-11-19(17)20;/h3-6,8-12H,7,13-15H2,1-2H3;1H

InChI key

KFYRPLNVJVHZGT-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Tricyclic antidepressant; inhibits the norepinephrine and serotonin transporters with Kis of 100 nM and 14.7 nM, respectively; high in vitro affinity for α1-adrenoceptors, serotonin and muscarinic acetylcholine receptors.

replaced by

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Repr. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S M Lee et al.
International journal of cardiology, 190, 353-359 (2015-05-06)
Amitriptyline (AMY) is a tricyclic anti-depressant that has recently been shown to have anti-inflammatory properties. We investigated whether AMY is cardioprotective against reperfusion injury in ex-vivo rat hearts. Thirty adult Sprague-Dawley rat hearts were perfused ex-vivo in a Langendorff apparatus.
Seth M Cohen et al.
Otolaryngology--head and neck surgery : official journal of American Academy of Otolaryngology-Head and Neck Surgery, 148(3), 374-382 (2013-01-10)
The goal of this systematic review was to examine the evidence for the use of the neuromodulating agents, amitriptyline, gabapentin, pregabalin, and baclofen, in the management of chronic, idiopathic cough patients. Online databases, including PubMed, Embase, Cochrane Review, and Web
Christian Janfelt et al.
Journal of mass spectrometry : JMS, 48(3), 361-366 (2013-03-16)
Displaced dual-mode imaging (DDI) is introduced as a method for simultaneous imaging in positive and negative-ion mode on the same sample with desorption electrospray ionization imaging, as well as a method for simultaneous imaging in full-scan and tandem mass spectrometry
Pascal Vanelderen et al.
Regional anesthesia and pain medicine, 38(2), 124-130 (2013-01-23)
In animal models of neuropathic pain (NP), promising results have been reported with the administration of minocycline, possibly through inhibition of spinal brain-derived neurotrophic factor (BDNF) expression. No data are available on the effect of amitriptyline and gabapentin on spinal
A M Agius et al.
Rhinology, 51(2), 143-153 (2013-05-15)
Patients often present to otolaryngologists with chronic facial pain, presumed to be of sinus origin despite normal nasal endoscopy and sinus CT. This pain has increasingly been recognized as being of neurological origin with one of the commonest underlying causes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service