Skip to Content
Merck
All Photos(1)

Key Documents

263532

Sigma-Aldrich

3,5-Difluoroaniline

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
F2C6H3NH2
CAS Number:
Molecular Weight:
129.11
Beilstein:
2802286
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

37-41 °C (lit.)

functional group

fluoro

SMILES string

Nc1cc(F)cc(F)c1

InChI

1S/C6H5F2N/c7-4-1-5(8)3-6(9)2-4/h1-3H,9H2

InChI key

KQOIBXZRCYFZSO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

In vitro nephrotoxic effects of 3,5-difluoroaniline has been investigated in renal cortical slices obtained from the kidneys of untreated, male Fischer 344 rats.

Application

3,5-Difluoroaniline has been used in the preparation of 3,5-difluorodimethylaniline.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

167.0 °F - closed cup

Flash Point(C)

75 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Further studies on the carcinogenicity of dyes related to 4-dimethylaminoazobenzene; the requirement for an unsubstituted 2-position.
J A MILLER et al.
Cancer research, 17(5), 387-398 (1957-06-01)
S K Hong et al.
Toxicology letters, 114(1-3), 125-133 (2000-03-14)
Haloanilines are widely used as chemical intermediates in the manufacture of pesticides, dyes and drugs. The purpose of this study was to examine the in vitro nephrotoxic effects of the four 4-haloaniline and four 3,5-dihaloaniline isomers using renal cortical slices

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service