Skip to Content
Merck
All Photos(1)

Key Documents

251674

Sigma-Aldrich

1-(3-Sulfopropyl)pyridinium hydroxide inner salt

98%

Synonym(s):

3-(1-Pyridinio)-1-propanesulfonate inner salt, PPS

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H11NO3S
CAS Number:
Molecular Weight:
201.24
Beilstein:
1621867
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

mp

280 °C (dec.) (lit.)

SMILES string

[O-]S(=O)(=O)CCC[n+]1ccccc1

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service