Skip to Content
Merck
All Photos(1)

Key Documents

246131

Sigma-Aldrich

7-Methoxy-4-methylcoumarin

99%

Synonym(s):

Methyl 4-methylumbelliferyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H10O3
CAS Number:
Molecular Weight:
190.20
Beilstein:
156900
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

99%

mp

158-160 °C (lit.)

SMILES string

COc1ccc2C(C)=CC(=O)Oc2c1

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maria E Riveiro et al.
Bioorganic & medicinal chemistry, 16(5), 2665-2675 (2007-12-07)
In the present study, we sought to establish the effect of diverse structural-related hydroxycoumarins on the proliferation, cytotoxicity, and induction of apoptosis in promonocytic leukemic cells (U-937). The dihydroxylated coumarins, 7,8-dihydroxy-coumarin and esculetin, induced DNA fragmentation as well as characteristic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service