Skip to Content
Merck
All Photos(1)

Key Documents

239798

1,3-Dioxolane

≥99.5%

Synonym(s):

Ethylene glycol methylene ether, Formaldehyde ethylene acetal

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H6O2
CAS Number:
Molecular Weight:
74.08
Beilstein:
102453
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

vapor density

2.6 (vs air)

vapor pressure

70 mmHg ( 20 °C)

Assay

≥99.5%

autoignition temp.

525 °F

contains

75 ppm BHT as inhibitor

refractive index

n20/D 1.401 (lit.)

bp

75-76 °C/1.013 hPa

mp

−95 °C (lit.)

density

1.06 g/mL at 25 °C (lit.)

SMILES string

C1COCO1

InChI

1S/C3H6O2/c1-2-5-3-4-1/h1-3H2

InChI key

WNXJIVFYUVYPPR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Charles A Ault et al.
Macromolecular rapid communications, 42(5), e2000692-e2000692 (2021-01-28)
Effects of molecular weight of methylphenyl-containing vinylsiloxy-functionalized terpolysiloxanes on their UV-activated crosslinking by hydrosilylation at room temperature in air, shelf life stability of "all-in-one" pastes prepared from them for additive manufacturing, and mechanical properties of the resulting crosslinked elastomers, are
Benito Alcaide et al.
The Journal of organic chemistry, 78(18), 8956-8965 (2013-09-07)
The gold-catalyzed preparation of 2-azetidinone-fused oxacycles was accomplished from β-lactam-linked enynes through heterocyclization reaction taking advantage of the acetonide pendant group. While the synthesis of fused tetrahydrofuran-β-lactams from 1,3-enynes could be considered as an unusual metal-catalyzed cyclization of enynols, α-alkoxy
Zhenhua Sun et al.
Nature communications, 8, 14627-14627 (2017-03-04)
Although the rechargeable lithium-sulfur battery is an advanced energy storage system, its practical implementation has been impeded by many issues, in particular the shuttle effect causing rapid capacity fade and low Coulombic efficiency. Herein, we report a conductive porous vanadium
Benjamin L Wegenhart et al.
Inorganic chemistry, 49(11), 4741-4743 (2010-05-06)
Low catalyst loading of a cationic oxorhenium(V) oxazoline complex, [2-(2'-hydroxyphenyl)-2-oxazolinato(-2)]oxorhenium(v), condenses diols and aldehydes to give 1,3-dioxolanes in excellent yields under neat conditions and reasonably mild temperatures. The reaction is applicable to biomass-derived furfural and glycerol. The resulting cyclic acetals
Matthias Schmidt et al.
Bioorganic & medicinal chemistry, 15(6), 2283-2297 (2007-02-06)
Multidrug resistance (MDR) to antitumor agents represents a major obstacle to a successful chemotherapy of cancer. Overexpression of P-glycoprotein (p-gp) seems to be the major factor responsible for MDR. A large number of chemically unrelated compounds are known to interact

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service