Skip to Content
Merck
All Photos(1)

Key Documents

234567

Sigma-Aldrich

Sodium fumarate dibasic

98%

Synonym(s):

Disodium fumarate, Fumaric acid disodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaOOCCH=CHCOONa
CAS Number:
Molecular Weight:
160.04
Beilstein:
4301302
EC Number:
MDL number:
UNSPSC Code:
12352100

Assay

98%

SMILES string

[Na+].[Na+].[O-]C(=O)\C=C\C([O-])=O

InChI

1S/C4H4O4.2Na/c5-3(6)1-2-4(7)8;;/h1-2H,(H,5,6)(H,7,8);;/q;2*+1/p-2/b2-1+;;

InChI key

MSJMDZAOKORVFC-SEPHDYHBSA-L

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service