Skip to Content
Merck
All Photos(1)

Documents

233935

Sigma-Aldrich

L-Malic acid disodium salt monohydrate

98%

Synonym(s):

(S)-(−)-Hydroxysuccinic acid disodium salt, (S)-(−)-Malic acid disodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaO2CCH2CH(OH)CO2Na · H2O
CAS Number:
Molecular Weight:
196.07
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:

Assay

98%

optical activity

[α]21/D −7.0°, c = 15 in H2O

SMILES string

O.[Na+].[Na+].O[C@@H](CC([O-])=O)C([O-])=O

replaced by

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service