Skip to Content
Merck
All Photos(1)

Key Documents

232556

Sigma-Aldrich

2-Chloro-3-ethylbenzoxazolium tetrafluoroborate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H9BClF4NO
CAS Number:
Molecular Weight:
269.43
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

mp

187-190 °C (lit.)

SMILES string

[F-].FB(F)F.CC[n+]1c(Cl)oc2ccccc12

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service