Skip to Content
Merck
All Photos(1)

Key Documents

228184

Sigma-Aldrich

Copper(II) cyclohexanebutyrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[C6H11(CH2)3CO2]2Cu
CAS Number:
Molecular Weight:
402.03
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
powder

grade

for analytical purposes

form

powder

technique(s)

AAS: suitable
liquid chromatography (LC): suitable

mp

126 °C (dec.) (lit.)

application(s)

battery manufacturing

SMILES string

O=C(CCCC1CCCCC1)O[Cu]OC(=O)CCCC2CCCCC2

InChI

1S/2C10H18O2.Cu/c2*11-10(12)8-4-7-9-5-2-1-3-6-9;/h2*9H,1-8H2,(H,11,12);/q;;+2/p-2

InChI key

FGUOYHAMMRMUQO-UHFFFAOYSA-L

Application

Solutions of copper(II) cyclohexanebutyrate can be used as calibration standards in the flame atomic absorption spectroscopy of copper foils from Li-ion batteries. [1] The salt was used in a study during which copper speciation in jet fuel was determined by liquid chromatography. [2]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Quantitation of the dissolution of battery-grade copper foils in lithium-ion battery electrolytes by flame atomic absorption spectroscopy.,
Zhao M, et al.
Electrochimica Acta, 49, 683?689-683?689 (2004)
Liquid chromatographic determination of copper speciation in jet fuel resulting from dissolved copper
Taylor DB and Synovec RE
Journal of Chromatography A, 659(1), 133-141 (1994)
H J Ougham et al.
Journal of bacteriology, 150(3), 1172-1182 (1982-06-01)
A strain of Arthrobacter was isolated by enrichment culture with cyclohexaneacetate as the sole source of carbon and grew with a doubling time of 4.2 h. In addition to growing with cyclohexaneacetate, the organism also grew with cyclohexanebutyrate at concentrations
Dean M Quesnel et al.
Chemosphere, 84(4), 504-511 (2011-04-05)
Naphthenic acids (NAs) are a major contributor to toxicity in tailings waste generated from bitumen production in the Athabasca Oil Sands region. While investigations have shown that bacteria can biodegrade NAs and reduce tailings toxicity, the potential of algae to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service