Skip to Content
Merck
All Photos(3)

Key Documents

223069

Sigma-Aldrich

4-Fluoro-N-methylaniline

97%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
234,00 kr.
5 G
416,00 kr.

234,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 G
234,00 kr.
5 G
416,00 kr.

About This Item

Linear Formula:
FC6H4NHCH3
CAS Number:
Molecular Weight:
125.14
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

234,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Assay

97%

refractive index

n20/D 1.5320 (lit.)

bp

79 °C/11 mmHg (lit.)

density

1.040 g/mL at 25 °C (lit.)

functional group

amine
fluoro

SMILES string

CNc1ccc(F)cc1

InChI

1S/C7H8FN/c1-9-7-4-2-6(8)3-5-7/h2-5,9H,1H3

InChI key

VLWRKVBQUANIGI-UHFFFAOYSA-N

Application

4-Fluoro-N-methylaniline was used as a model compound to study the in vivo and in vitro biotransformation of secondary aromatic amines[1].

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

168.8 °F - closed cup

Flash Point(C)

76 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M G Boersma et al.
Drug metabolism and disposition: the biological fate of chemicals, 21(2), 218-230 (1993-03-01)
In vivo and in vitro biotransformation of secondary aromatic amines was investigated using 4-fluoro-N-methylaniline as the model compound. Attention was focused on the role of cytochromes P-450 and the flavin-containing monooxygenase in formation of the various metabolic products. In vitro
James P Driscoll et al.
Chemical research in toxicology, 23(5), 861-863 (2010-04-08)
Here, we report on the mechanism by which flavin-containing monooxygenase 1 (FMO1) mediates the formation of a reactive intermediate of 4-fluoro-N-methylaniline. FMO1 catalyzed a carbon oxidation reaction coupled with defluorination that led to the formation of 4-N-methylaminophenol, which was a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service