Skip to Content
Merck
All Photos(1)

Key Documents

222909

Sigma-Aldrich

N-Phenylthiourea

97%

Synonym(s):

1-Phenyl-2-thiourea, PTU, Phenylthiocarbamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5NHCSNH2
CAS Number:
Molecular Weight:
152.22
Beilstein:
907309
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

mp

145-150 °C (lit.)

SMILES string

NC(=S)Nc1ccccc1

InChI

1S/C7H8N2S/c8-7(10)9-6-4-2-1-3-5-6/h1-5H,(H3,8,9,10)

InChI key

FULZLIGZKMKICU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Oral - Skin Sens. 1

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anna A Brozyna et al.
International journal of cancer, 123(6), 1448-1456 (2008-06-21)
Melanin pigment displays strong photo- and radioprotective properties, suggesting that inhibition melanogenesis could increase sensitivity of melanoma to ionizing radiation. We tested this concept in human melanoma cells cultured in either Ham's F10 medium to maintain amelanotic phenotype or DMEM
Xiangguo Shi et al.
Blood, 125(19), 2974-2984 (2015-03-18)
Isocitrate dehydrogenase 1 mutation (IDH1-R132H) was recently identified in acute myeloid leukemia with normal cytogenetics. The mutant enzyme is thought to convert α-ketoglutarate to the pathogenic 2-hydroxyglutarate (2-HG) that affects DNA methylation via inhibition of ten-eleven translocation 2. However, the
Warrick J Brewer et al.
Psychiatry research, 199(1), 8-11 (2012-04-17)
Reports suggesting that schizophrenia participants are more likely to be phenylthiocarbamide (PTC) non-tasters when compared to controls have recently been controversial. If supported, a genetic-based phenotypic variation in PTC taster status is implicated, suggesting a greater illness risk for those
Laila Gannoun-Zaki et al.
Antimicrobial agents and chemotherapy, 57(1), 629-632 (2012-11-02)
The mechanism by which the antitubercular drug isoxyl (ISO) inhibits mycolic acid biosynthesis has not yet been reported. We found that point mutations in either the HadA or HadC component of the type II fatty acid synthase (FAS-II) are associated
Thomas Grau et al.
Antimicrobial agents and chemotherapy, 56(2), 1142-1145 (2011-11-23)
Ethionamide (ETH) is a second-line drug for the treatment of tuberculosis. As a prodrug, ETH has to be activated by EthA. ethA is controlled by its repressor EthR. 2-Phenylethyl-butyrate (2-PEB) inhibits EthR binding, enhances expression of EthA, and thereby enhances

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service