21885
γ-Carboxy-DL-glutamic acid
≥99.0%
Synonym(s):
3-Amino-1,1,3-propanetricarboxylic acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
NH2CH(COOH)CH2CH(COOH)2
CAS Number:
Molecular Weight:
191.14
Beilstein:
2214048
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
≥99.0%
reaction suitability
reaction type: solution phase peptide synthesis
application(s)
peptide synthesis
storage temp.
2-8°C
SMILES string
NC(CC(C(O)=O)C(O)=O)C(O)=O
InChI
1S/C6H9NO6/c7-3(6(12)13)1-2(4(8)9)5(10)11/h2-3H,1,7H2,(H,8,9)(H,10,11)(H,12,13)
InChI key
UHBYWPGGCSDKFX-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Packaging
Bottomless glass bottle. Contents are inside inserted fused cone.
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service