Skip to Content
Merck
All Photos(1)

Key Documents

217530

Sigma-Aldrich

7-Ethoxy-6-methoxy-2,2-dimethylchromene

99%

Synonym(s):

Ethoxy-Precocene, Ethoxyprecocene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H18O3
CAS Number:
Molecular Weight:
234.29
EC Number:
MDL number:
UNSPSC Code:
12352103

Assay

99%

SMILES string

CCOc1cc2OC(C)(C)C=Cc2cc1OC

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Azambuja et al.
Brazilian journal of medical and biological research = Revista brasileira de pesquisas medicas e biologicas, 20(2), 175-179 (1987-01-01)
1. Topical application of the proallatotoxin ethoxyprecocene II (60 micrograms/nymph) to 4th-instar nymphs of Rhodnius prolixus at various times before and after a blood meal induced precocious metamorphosis. 2. The highest percentage of adultoids was obtained when proallatotoxin was applied
Irfan Ahmad Khan et al.
Indian journal of experimental biology, 41(4), 328-335 (2004-07-17)
Administration of a synthetic precocene analogue, 7-ethoxy-6-methoxy-2,2-dimethyl chromene to fourth and last instar larvae of E. vitella results into heterochrony, viz. prothetely and metathetely. These disturbances are due to interference with the endocrine system and application of juvenile hormone to
E S Garcia et al.
Brazilian journal of medical and biological research = Revista brasileira de pesquisas medicas e biologicas, 18(2), 195-199 (1985-01-01)
Oogenesis in R. prolixus females is induced by feeding blood, blood plasma, a suspension of washed erythrocytes, or 5% solutions of egg albumin, bovine serum albumin or hemoglobin. Unfed females and those fed saline, casein hydrolysate, dextran or sucrose do
G Gellissen et al.
Canadian journal of biochemistry, 59(8), 648-654 (1981-08-01)
The lipid-transport lipoprotein, lipophorin (Lp), an abundant plasma protein of insects, is synthesized in the fat body of Locusta migratoria as a polypeptide of relative mass (Mr) = 85 000. No posttranslational modification could be detected. Precipitation of this protein
In vitro inactivation of tenebrio molitor corpora allata by a synthetic precocene analog.
W S Bowers et al.
General and comparative endocrinology, 47(1), 120-124 (1982-05-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service