Skip to Content
Merck
All Photos(1)

Key Documents

202169

Sigma-Aldrich

Chromium(III) oxide

powder, 1 μm, ≥98%

Synonym(s):

Chromia

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Cr2O3
CAS Number:
Molecular Weight:
151.99
EC Number:
MDL number:
UNSPSC Code:
12352300

Assay

≥98%

form

powder

particle size

1 μm

SMILES string

O=[Cr]O[Cr]=O

InChI

1S/2Cr.3O

InChI key

QDOXWKRWXJOMAK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Used in a solid-state reaction to form the new ternary oxide, CrVMoO7, which should find application in selective oxidation. Its crystal structure was also determined.
Used in a solid-state reaction to form the new ternary oxide, CrVMoO7, can be used in selective oxidation.

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wang, X. et al.
Inorganic Chemistry, 37, 3252-3252 (1998)
Poornima Vajpayee et al.
Journal of biomedical nanotechnology, 7(1), 205-206 (2011-04-14)
In this study, seeds of Triticum aestivum L. (Poaceae) were exposed to 0-100 microg/mL chromium oxide nanoparticles (Cr2O3, Nps) to study the phytotoxic effects on seed germination and seedling growth. It has been observed that 25-100 microg/mL Cr2O3, Nps inhibited
Nien-Hsun Li et al.
Journal of hazardous materials, 198, 356-361 (2011-11-15)
The feasibility of stabilizing nickel-laden sludge by a high-temperature NiCr(2)O(4) synthesis process was investigated with different sintering temperatures, salt contents, molar ratios, and reaction atmospheres. The crystalline phases of species were investigated by using an X-ray diffraction, and the surface
S E Dutton et al.
Journal of physics. Condensed matter : an Institute of Physics journal, 23(24), 246005-246005 (2011-06-01)
α-SrCr(2)O(4) has a triangular planar lattice of d(3) Cr(3+) made from edge sharing CrO(6) octahedra; the plane shows a very small orthorhombic distortion from hexagonal symmetry. With a Weiss temperature of - 596 K and a three-dimensional magnetic ordering temperature
Isabella Marr et al.
Sensors (Basel, Switzerland), 11(8), 7736-7748 (2011-12-14)
Zeolites are promising materials in the field of gas sensors. In this technology-oriented paper, a planar setup for potentiometric hydrocarbon and hydrogen gas sensors using zeolites as ionic sodium conductors is presented, in which the Pt-loaded Na-ZSM-5 zeolite is applied

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service